Would you like to log in again?
Accredo Safety Lockout Tagout Hasp 24 Hole, ZING Enterprises
Supplier: ZING Green Safety Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7241
76026-320EA 42.59 USD InStock
76026-320
Accredo Safety Lockout Tagout Hasp 24 Hole, ZING Enterprises
Security Locks
Lockout tagout hasp.
  • Supports OSHA Standard 1910.147
  • High quality
  • Very durable
24 Hole Lockout Hasp is designed to secure switches, valves and other energy devices from use while equipment is being serviced or repaired. The hasp is made from durable coated steel and measures 14’’L x 2.25’’W. The closed hasp hole size measures 1 x 1 ½’’. The hasp will accommodate 24 padlocks with a max shackle diameter of 3/8’’.
Certifications: OSHA 1910.147
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty