Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accredo Safety Squeezer Multipurpose Cable Lockout, ZING Enterprises

Supplier: ZING Green Safety Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7244 7245
76026-326EA 30.41 USD InStock
76026-326 76026-328
Accredo Safety Squeezer Multipurpose Cable Lockout, ZING Enterprises
Security Locks
High quality lockout device.

  • High quality
  • Effective lockout device
  • Very durable

Multi-purpose cable lockout features a durable plastic body and a 3 ft

Also available with a 3 ft. di-electric nylon cable.

steel cable. The lockout body measures 1D x 6L x 2W and will accommodate up to 6 padlocks with shackles up to ¼. The lockout is easy to install: simply route the cable to lockout the device then feed the cable down the center point at the top. When the cable is tight and the source is locked out, squeeze the handle and apply padlock.

Certifications: OSHA 1910.147

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty