Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accredo Safety Ball Valve Lockout, ZING Enterprises

Supplier: ZING Green Safety Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7251 7249 7250
76026-336EA 41.64 USD InStock
76026-336 76026-340 76026-338
Accredo Safety Ball Valve Lockout, ZING Enterprises
Security Locks
Convenient way to lockout ball valves.

  • High quality
  • Effective lockout device
  • Very durable

Small Ball Valve Lockout, is made from non-conductive, durable plastic (polyurethane), and fits pipe sizes 3/8" to 1-1/4", and handle sizes 3/8" to 1-1/4".

Ball Valve Lockout, is made from non-conductive, durable plastic (polyurethane), and fits pipe sizes 1-1/2" to 2-1/2", and handle sizes 1-1/2" to 2-1/2".

Large Ball Valve Lockout, is made from non-conductive, durable plastic (polyurethane), and fits pipe sizes 2" to 8", and handle sizes 2" to 8".

Certifications: OSHA 1910.147

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty