Would you like to log in again?
Accredo Safety Oversize Push Button Lockout, ZING Enterprises
Supplier: ZING Green Safety Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7254
76026-346EA 38.54 USD InStock
76026-346
Accredo Safety Oversize Push Button Lockout, ZING Enterprises
Security Locks
Supports OSHA Standard 1910.147.
  • Durable Acrylic Construction
  • High quality
  • Safety device
Oversize Push Button Lockout provides a safe and effective method for locking out emergency push buttons. The lockout is made from durable plastic (acrylic) and measures 3.5’’H x 5’’L x 3.2’’W, with a 1 4/5’’ diameter hole in the center. The lockout accommodates 1 padlock with a maximum shackle diameter of ¼’’.
Certifications: OSHA 1910.147
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty