Would you like to log in again?
Accredo Safety Slider Pin Circuit Breaker Lockout, ZING Enterprises
Supplier: ZING Green Safety Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7261 7260 7262 7259
76026-358EA 18.38 USD InStock
76026-358 76026-356 76026-362 76026-360
Accredo Safety Slider Pin Circuit Breaker Lockout, ZING Enterprises
Security Locks
A variety of sizes and configurations are available.
  • No Tools Required for Installation
  • Effective lockout device
  • High quality
120/277 Miniature Circuit Breaker Lockout - Pin In, Standard, provides an effective lockout solution
120/277 Miniature Circuit Breaker Lockout - Pin Out, Standard, provides an effective lockout solution. The lockout measures 3/4"H, 2-3/4''L, 3/4"W and accommodates 1 padlock, with a maximum shackle diameter of 9/32''. The lockout is constructed from durable plastic and is designed for use with ISO/DIN miniature breakers. No tools are required for installation.
120/277 Miniature Circuit Breaker Lockout - Pin Out, Wide, provides an effective lockout solution. The lockout measures 3/4"H, 2-3/4''L, 3/4"W and accommodates 1 padlock, with a maximum shackle diameter of 9/32''. The lockout is constructed from durable plastic and is designed for use with ISO/DIN miniature breakers. No tools are required for installation.
Miniature Circuit Breaker Lockout Set includes 3 miniature circuit breaker lockouts: Pin In Standard, Pin Out Standard, and Pin Out Wide. The lockouts measure 3/4"H, 2-3/4''L, 3/4"W and accommodate 1 padlock each, with a maximum shackle diameter of 9/32''. The lockouts are constructed from durable plastic and are designed for use with ISO/DIN miniature breakers. No tools are required for installation.
Certifications: OSHA 1910.147
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty