Would you like to log in again?
Accredo Safety Pin And Sleeve - Socket Lockout, ZING Enterprises
Supplier: ZING Green Safety Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7267 7265 7264 7266
76026-368EA 49.87 USD InStock
76026-368 76026-366 76026-372 76026-370
Accredo Safety Pin And Sleeve - Socket Lockout, ZING Enterprises
Security Locks
A variety of sizes and configurations are available.
  • High quality
  • Effective lockout device
  • Very durable
Pin and Sleeve Socket Lockout, Small, prevents electrical plugs from being inserted into sockets
Pin and Sleeve Socket Lockout, Medium, prevents electrical plugs from being inserted into sockets. The device is for use with plugs from 220-240V.
Pin and Sleeve Socket Lockout, Large, prevents electrical plugs from being inserted into sockets. The device is for use with plugs from 380-415V.
Extra Large Pin and Sleeve Socket Lockout prevents electrical plugs from being inserted into sockets. The device is for use with 500V plugs.
Certifications: OSHA 1910.147
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty