Would you like to log in again?
Accredo Safety Group Lock Box, ZING Enterprises
Supplier: ZING Green Safety Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7288 7286
76026-386EA 105.36 USD InStock
76026-386 76026-390
Accredo Safety Group Lock Box, ZING Enterprises
Security Locks
High quality lockout box.
  • High quality
  • Group lockout purposes
  • Very durable
Group Lockout Box, Steel, protects workers in group maintenance and repair operations
Wall-Mountable Group Lockout Box protects workers during maintenance and repair operations. Accommodates up to 12 padlocks, plus 1 on the latch.
The box accommodates up to 7 padlocks.
Certifications: OSHA 1910.147
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty