Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VIAL URINALYSIS W/YELLOW CAP PP AMB 12ML
Supplier: CAPITOL VIAL INC.
Urinalysis Vial 12mL^ Urinalysis Vial, Color: Amber, with Yellow Cap with Preservative pill, Material: Polypropylene, Stabilize urine specimens for up to 72 hours, Size: 12 mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL URINALYSIS W/YELLOW CAP PP AMB 12ML 76026-426 S40824 Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty