Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Chex-All® III Instant-Seal Pouches, Propper Manufacturing
Supplier: Propper
Chex-All®
02431800 02431000 02431300 02430900 02431400
76026-466CS 211.43 USD InStock
76026-466 76026-476 76026-474 76026-472 76026-470
Chex-All® III Instant-Seal Pouches, Propper Manufacturing
Bags Sterilization Pouches
Chex-All® III self-seal sterilization pouches for use in steam autoclaves or ethylene oxide sterilizers provide a barrier to maintain sterility after the sterilization process.
  • Self-sealing serilization pouches for use in steam and ethylene oxide
  • Pre-printed indicator inside and outside the pouch
  • Features triple-sealed paper/plastic film to ensure the security of contents
  • Made of 2.5 mm plastic and 73 to 74 gram paper
  • Chemical indicator provides distinct color change from yellow to black in steam
  • Chemical indicator provides distinct color change from red to green in EO
Chex-All® III pouches offer breathability, a sterile barrier, and the advantage of pre-printed chemical indicators (both inside and outside the pouch) along with the convenience of self-sealing pouches. ³/₄" wide adhesive line guarantees a strong seal that will not be affected by steam or ethylene oxide exposure. Chex-All® III pouches are for use in steam and ethylene oxide, eliminating the need for two sets of pouches. Chex-All pouches are validated for double pouch packaging.
Certifications: Meets ANSI/AAMI Standard ST-79.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty