Would you like to log in again?
BCR® Foam Flat Mop Cover, Berkshire
Supplier: Berkshire
BCR®
FMC6LF12
76028-246CS 767.92 USD InStock
76028-246
BCR® Foam Flat Mop Cover, Berkshire
Mops
Abrasion-resistant, polyester-knit cleanroom fabric laminated to absorbent polyurethane foam.
  • Abrasion resistant
  • Laser cut
  • High absorbency
The V Flat Mop Foam Cover is laser cut and cleanroom laundered to minimize fiber and particle release
This mop cover is compatible with a wide range of chemicals, disinfectants, and cleaners. It is autoclavable and compatible with both gamma and ETO sterilization.
The Flat Mop Foam Cover is abrasion-resistant, and constructed of polyester-knit cleanroom fabric which is laminated to absorbent polyurethane foam. The 10 mm thickness provides a cushioned surface for optimum conformance to uneven floors, walls, and ceilings.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty