Would you like to log in again?
CUNO™ CTG-Klean System Filter Pack with Betapure™ AU Series Filter Cartridge
Supplier: 3M Healthcare
CUNO™ 3M™
7000050978 7010314611 7010380772 7010314609 7010339919 7010339554 7010302970 7010379381 7010380768 7010338819 7010379389 7100113917 7010380746 7010338816 7010338429 7010292986 7010379387 7100025230 7010380744 7010338839 7010379383 7010380745 7010380775 7010379393 7010379392 7100113918 7010338813 7010314602 7010380766 7010302965 7000050459 7010379382 7010295923 7010314598 7010338815 7010380774 7010380881 7010338817 7010338838 7010415696 7010338834 7010380773 7010380767 7010379394
76028-861CS 1425.43 USD InStock
76028-861 76029-325 76029-247 76029-249 76029-329 76029-295 76029-251 76029-253 76028-899 76028-897 76028-875 76028-853 76029-291 76028-859 76029-293 76028-857 76031-781 76028-891 76028-873 76028-851 76028-895 76028-893 76028-871 76029-317 76029-319 76031-759 76029-283 76029-285 76029-241 76029-321 76031-777 76029-243 76029-287 76029-323 76031-757 76031-779 76029-289 76029-245 76028-889 76031-773 76031-753 76031-775 76028-901 76029-281
CUNO™ CTG-Klean System Filter Pack with Betapure™ AU Series Filter Cartridge
Filters Liquid Process Filters
CUNO™ CTG-Klean system filter pack with Betapure™ AU series cartridge as part of a totally enclosed system that uses a separate pressure vessel and disposable filter pack to isolate the product from the housing. It reduces the extensive time, labor and operator exposure to impurities and cleaning solvents involved with filter change-outs and cleaning of conventional housings.
  • Cartridge pack design reduces the extensive time and labor involved with filter change-outs and cleaning of conventional housings
  • Totally enclosed filter pack avoids handling of used filter cartridges
  • Design reduces gaskets, posts, spring and seal assemblies, and seal plates for efficient, cost- effective filtration
  • Filter pack can be capped and reused, depending on the application
  • Rapid filter change-out reduces labor time and the need for duplex systems
  • Betapure AU series filter cartridge’s rigid structure provides consistent and reproducible contaminant reduction
  • Betapure AU series filter cartridge provides excellent reduction of deformable contaminants for consistent effluent quality
CUNO™ CTG-Klean System was designed with versatility in mind to accommodate a wide variety of filter media, and offers compatibility with a range of fluids to meet a variety of process requirements.
Our line of Betapure™ AU series filters are state-of-the-art rigid, bicomponent fiber depth filters, available in absolute ratings from 2 to 190 μm. We manufacture them using our advanced fiber technology to provide the ultimate in consistent classifying filtration performance. The bonded rigid structure is non-deformable to withstand high differential pressure, fiber shedding, and to reduce media migration and provide absolute filtration - batch to batch. Betapure™ AU Series Filter Applications include Coatings - high quality paint, film coatings, resins, and ink, Food and beverage - bottled water, Pre-Ro, blend water, wash water, General industrial - desalination, plating, machine tool coolant, process water, Electronics - Pre-Ro, wafer slurries, ceramic slurries, chemical mechanical polishing, cathode ray tube production, disc cleaning and Chemical/petro-chemical - process water, Pre-Ro, amine, fine chemicals.
The rigid construction advantage.
To meet demanding filtration quality standards in today's market, absolute ratings will provide you with product consistency, and improve yields.
Lower pressure drops mean increased filter life, product throughput, and permit the use of fewer filters to achieve a given flow vs. differential pressure. When appropriately sized and rated, Betapure AU series filter cartridges employed in the CTG-Klean™ system work to meet your application specifications.
Go with Our Flow
Our cartridge pack design reduces the extensive time and labor involved with filter change-outs and cleaning of conventional housings, thereby reducing operator exposure to product cleaning solvents. This streamlined system, coupled with high performance, long-life filters, make CUNO CTG-Klean Systems the clear choice for cost-effective filtration.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty