Would you like to log in again?
CUNO™ CTG-Klean System Filter Hardware and Accessories
Supplier: 3M Healthcare
7010301280 7010415652 7010295959 7010379818 7010382135 7010339955 7010338241 7100336131 4010046396 7010339681
76029-000CS 11074.37 USD InStock
76029-000 76029-010 76029-002 76029-012 76029-006 76029-204 76028-998 76029-348 76028-996
CUNO™ CTG-Klean System Filter Hardware and Accessories
Filters Liquid Process Filters
CUNO™ CTG-Klean systems filter hardware and accessories with all of the components needed for installation, set-up and filter pack replacement. CUNO CTG-Klean systems hardware and accessories simplify system installation and give you flexibility. All components are manufactured from stainless steel for extended service life, exceptional durability, and ease of cleaning.
  • Hardware and accessories help extend the capabilities of the CUNO™ CTG-Klean System
  • Easy-to-fit components help save time and labor costs
  • Stainless steel construction for long service life, durability, and ease of cleaning
  • Allows the use of shorter filter packs for reduced total filtration cost
Flexible Line Plumbing Kit has inlet and outlet ports on the housing head, which must be reduced to allow access for installation and for removal of the pack. For optimal performance, high pressure stainless steel flexible plumbing should be used instead of 'hard' piping. We offer a standard plumbing kit that includes the threaded hose, ball-valves, all connectors, and pressure gauges to connect with no need for additional parts.
System Flushing Elbow - Cleaning conventional filter housings requires solvent use. With the CUNO™ CTG-Klean system, the process liquid does not come in contact with the housing, so it is only necessary to flush the inlet and outlet plumbing. These lines, connected by the flushing elbow, can be flushed with the CUNO™ CTG-Klean system off-line. The stainless steel flushing elbow is available for all CUNO™ CTGKlean system filter housing configurations and is supplied with one or two fluorocarbon O-rings.
In processes where the batch size can vary or where process changes reduce the contaminant load, fewer filter cartridges could be used if a smaller housing were available. The CUNO™ CTGKlean system Volume Reducer drives down the cost of filtration by allowing the use of shorter filter packs to cut the total filtration cost. The volume reducer is available for the 3WTS and 7WTS configurations to allow use of a single high filter pack within the two-high housing (3WTS2 and 7WTS2), or a two-high filter pack in the three-high housing (3WTS3 and 7WTS3).
Tilting Device
Our tilting mechanism provides greater ease of use and the recovery of up to 95% of the hold-up volume from the filter pack. It also helps in lifting the potentially heavy used filter pack for ease of removal from the housing. And our durable stainless steel construction makes the tilting device ideal for stand-alone or skid mounted service.
CUNO™ CTG-Klean System Housing Dimensions
The CUNO™ CTG Klean System housings are available in one-around, three-around, and seven-around sizes, with varying filter pack heights. The 3M™ 1WTS is a one-around housing that is configurable to hold a 10" or 20" cartridge filter pack lengths. The three-around 3WTS housings can be configured to hold either a 10", 20", or 30" cartridge. The largest housing is the 7WTS, which is a seven-around housing that is configurable to 10", 20" or 30" cartridge length filter pack.
Our complete CUNO™ CTG-Klean system design is engineered to minimize operator exposure to the fluid and reduce the time and labor required for cleaning, and our line of hardware and accessories extend the capabilities of the system while preserving these cost-saving features.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty