Would you like to log in again?
3M™ Zeta Plus™ BC Series Filter Capsule with SP Series Media
Supplier: 3M Healthcare
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Zeta Plus™
7000051189 7100103022 7000148476 7010338681 7100002302 7000051190 7100000582 7010338578 7100004090 7010338558 7010338564 7000125771 7010380632 7010338576 7010303169 7000001817 7000051184 BC0025L60SP02A 7100005501 7000125767 7000051188 7010382088 7010382086 7100107846 7010409445 7000125768 7100004078 7010338560 7010382104 7010382090 7000001819 7000148477
76032-129CS 606.41 USD InStock
76032-129 76029-607 76029-629 76029-609 76029-567 76029-643 76029-547 76029-623 76029-603 76029-605 76029-627 76029-551 76029-554 76029-631 76029-553 76032-139 76032-137 76029-633 76029-611 76032-131 76029-555 76029-613 76029-557 76029-615 76032-135 76029-537 76029-559 76029-639 76029-617 76029-539 76029-541 76029-641
3M™ Zeta Plus™ BC Series Filter Capsule with SP Series Media
Filters Capsule Filters
Zeta Plus BC Series Filter Capsules with SP Series Media for biological, small molecule pharmaceutical, and food and beverage manufacturing applications where an optimized balance of throughput and particle removal efficiency is critical. These capsules, having the same media as production products, are ideal for assessing filtration media grades using low challenge fluid volumes.
  • Zeta Plus SP Series filter media is offered in the largest selection of grades in the 3M depth filter portfolio with a range of porosity and tightness suitable for diverse applications from cell culture harvest to fine clarification
  • Single layer media configurations are available in capsule and cartridge formats for high surface loading filtration applications
  • BC25 Laboratory capsules contain same media as in production scale systems and are ideal for selecting the optimum media grade using low challenge fluid volume
  • Full range of scalable single-use capsules and cartridges enables predictable performance from the laboratory to commercial production
Zeta Plus SP Series filter media is a mixture of high surface area inorganic particles, cellulose and a positively charged crosslinking polymer.
The combination of filtration efficiency and throughput results from the balance between media porosity, depth, and the relatively low level anion exchange capacity: removing some negatively charged soluble contaminants and sub-micron particles via anion exchange and larger particles by mechanical sieving. Single layer formats are offered in our highest filter surface area, making them the preferred choice over double layer formats for hard particle removal and predominantly surface loading filtration applications. The Zeta Plus™ BC Series Filter Capsule with SP Series media is ideal for optimizing the filtration performance for each drug process by effectively screening the breadth of available media options. A scalable range of single-use capsules and cartridges enables predictable performance at every stage of drug development from early stage discovery to commercial production.
Pharmaceutical Grade Product.
The Zeta Plus™ BC Series Filter Capsules with SP Series Media are designed for use in the manufacture of biopharmaceutical, vaccine, blood fractionation, small molecule API drug substances, and food and beverages. The effluent characteristics of the Zeta Plus SP Series Filter media, capsules and cartridges have been tested following a number of protocols contained in the United States Pharmacopeia (USP) compendia.
Filtration Performance
Zeta Plus SP Series filter media is a mixture of high surface area inorganic particles, high surface area cellulose and a crosslinking polymer resin. The polymer resin has a positive charge as a result of a unique balance of secondary, tertiary and quaternary amines resulting in a relatively low anion exchange capacity. As such, Zeta Plus™ BC Series Filter Capsules with SP Series Media are a good choice in applications where the target product is negatively charged and product recovery may be of concern. By controlling the choice of raw materials and the formulation, a very large selection of filtration grades are produced, each with a unique balance of fine particle removal and throughput capacity.
Scalable Single-use Capsules
A full range of scalable filter product configurations enables predictable performance at every stage of drug development from early stage discovery to commercial production. Specifically, the 3M™ Encapsulated System enables a completely single‐use solution to process from 0.5 L to 2500+ L of liquid. The BC25 laboratory capsule with 25 cm²; filter media is ideal for media grade screening using small liquid volumes. The scale‐up capsules with 170; 340 and 1020 cm²; media are designed for scale‐up studies and low volume production. Production capsules having 0.23 m² and 2.5 m² single layer media complete the single-use capsule line.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty