Would you like to log in again?
3M™ CUNO Legacy Drop-In Style Replacement Cartridge for Ice Maker Applications, Model CFS420IMF, 5560905
Supplier: 3M Healthcare
3M™
7100040600 5560905
76030-081CS 4438.27 USD InStock
76030-081 76030-080
3M™ CUNO Legacy Drop-In Style Replacement Cartridge for Ice Maker Applications, Model CFS420IMF, 5560905
Filters Filter Cartridges
3M™ CUNO legacy drop–in style replacement cartridge for ice maker applications, Model CFS420IMF, when used in a drop–in style filtration housing, provides consistent quality water by reducing particulate. The built–in scale inhibitor helps to reduce potential harmful effects of scale build–up on equipment.
  • 9³/₄" drop–in style replacement cartridge
  • Provides consistent quality water
  • Reduces particulate from incoming water
  • Graded density design built–in scale inhibitor helps to reduce potential harmful effects of scale build–up on equipment
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty