Would you like to log in again?
3M™ Micro-Klean™ D Series Filter Cartridge
Supplier: 3M Healthcare
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Micro-Klean™
7010339332 7100010279 7000001621 7000001695 7010314623 7000001618 7000125459 7100139437 7010303114 7100003995 7000125357 7100026654 7010380550 7000125356 7100064081 7100074376 7100064775 7000148448 7010314093 7100031967 7010380908 7010302978 7100006302 7010380048 7100003470 7000001694 7000001696 7010409018 7100008957 7100140743 7010381681 7000050475 7000125513 7000125452 7010338866 7010338655 7000125460 7000050498 7010338660 7010338867 7010314807 7100008196 7100008836 7010339315 7100031479 7010314094 7100085774 7010338865 7010382809 7100008207 7000050477 7000050269 7100001253 7010380900 7100008817 7010338183 7100026699 7010314084 7100087212 7010339719 7010380940 7000125451 7000125450 7010380814 7100098564 7100064032 7100083754 7010338862 7010338868 7010380905 7000125349 7100139416 7100059113 7000001620 7000125454 7010380936 7010338860 7100001254 7000125455 7010295966 7100066716 7010380060 7100065014 7100064030 7000029251 7100001252 7010380907 7010381145 7010409688 7100067774 7000125457 7000125461 7100139439 7000027564 7100031510 7100064031 7010412804 7010315150 7000050236 7100055078 7100056058
76033-171CS 667.6 USD InStock
76033-171 76030-117 76030-753 76030-751 76030-113 76030-111 76030-197 76030-193 76033-217 76030-191 76033-173 76033-177 76033-179 76033-211 76030-129 76033-181 76032-873 76030-123 76030-121 76033-183 76033-185 76033-189 76030-139 76030-137 76030-135 76030-133 76030-099 76030-131 76033-159 76030-097 76030-093 76033-151 76033-155 76033-157 76030-149 76030-147 76030-145 76033-203 76033-209 76033-161 76033-163 76033-165 76033-201 76032-841 76032-847 76030-159 76030-157 76030-155 76032-289 76030-153 76030-151 76032-839 76032-833 76030-729 76030-727 76032-831 76032-837 76030-725 76030-203 76030-169 76030-167 76030-163 76030-161 76033-669 76030-739 76030-737 76032-863 76033-191 76030-735 76032-869 76033-193 76032-867 76030-731 76030-179 76030-173 76033-635 76033-637 76033-195 76033-197 76028-573 76032-853 76030-749 76030-747 76032-859 76030-109 76032-857 76030-745 76033-639 76030-741 76030-189 76030-187 76031-539 76033-643 76030-185 76033-645 76030-183 76030-181 76033-647 76032-291 76032-849 76033-641
3M™ Micro-Klean™ D Series Filter Cartridge
Filters Filter Cartridges
Micro-Klean™ D Series filter cartridge utilizing blanketed filter technology that combines an enhanced open-wind process with an internal media blanket to achieve superior flow rates, filtration efficiency, and consistent filtration characteristics from cartridge to cartridge, lot to lot.
  • Cotton materials and polypropylene twine/polypropylene media are FDA 21 CFR listed materials and suitable for food and beverage
  • Graded density construction prevents significant pressure drops
  • Integral lengths from 9⁷/₈" to 40" reduces joints that cause blinding or by-pass
The performance of the Micro-Klean™ D Series Filter Cartridge is directly related to the advanced winding pattern of the yarn matrix combined with our process of separately applying a tailored media 'blanket' between successive layers of yarn.
Micro-Klean D Series winding pattern also provides less restriction than the patterns common to ordinary wound cartridges. Consequently, the Micro-Klean D Series Filter Cartridge provides up to ²/₁₂ times more open area, enhancing the flow rate by up to 500% for the same pressure drop. Micro-Klean D Series Filter Cartridge is a blanket media cartridge offering true graded density, with more open filtration on the outside of the filter and fine, efficient filtration on the inner layer of the cartridge. Clogging of the filtration surfaces by large particles is minimized and cartridge life extended which can lower cartridge replacement costs.
Micro-Klean D Series Filter Cartridges are nominally rated from 0.5 to 350 µm and are available with various media, matrix, and core materials for compatibility within different processes. Standard materials include cotton yarn/cotton media blanket for use in applications involving water, alcohol, and other polar liquids. The cotton materials and PP twine/PP media we use are FDA 21 CFR listed materials for use with potable water, food, and beverage products. Cartridges can be configured with tinned steel, stainless steel, or polypropylene cores. The use of various core materials gives you an advanced range of compatibility. Micro-Klean D Series Filter Cartridges can also be configured with various end-treatments and O-ring materials to fit competitive filter housings.
This winding pattern creates much larger diamond shaped contaminant holding chambers. The separately inserted blanket encloses the chambers and maintains the consistency and integrity of filtration. These two factors combine to achieve a balance of filtration characteristics, which are hard to obtain with ordinary wound filter cartridges that use teased or brushed fibers on the yarn to produce a filtering media.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty