Would you like to log in again?
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 20 Gal. Drawdown Tank w/Connection Kit, 5598408
Supplier: 3M Healthcare
3M™
7000029227
76030-244EA 3519.74 USD InStock
76030-244
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 20 Gal. Drawdown Tank w/Connection Kit, 5598408
Tanks Cylindrical Tanks
3M™ commercial reverse osmosis water storage tanks, model 20 gal. Drawdown tank with connection kit, 5598408 is appropriately sized for boiler-based steamer units up to 300000 BTUs.
  • Reverse osmosis 20 gallon drawdown tank at 60 psi with 15 psi pre-charge
  • Polyethylene tank liner and fiberglass wrap tank exterior
Drawdown tank with connection kit, 5598408 contains a polyethylene tank liner and fiberglass wrap tank exterior. Tanks can be plumbed together to accommodate larger steamer operations. Fittings are designed to accept plumbing from 3M™ reverse osmosis systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty