Would you like to log in again?
3M™ Valve-In-Head/Bracket Assembly, Model VH3-NPT, 6213033, 3M
Supplier: 3M
3M™
7100163109
76030-649CS 943.01 USD InStock
76030-649
3M™ Valve-In-Head/Bracket Assembly, Model VH3-NPT, 6213033, 3M
Valves Vacuum Valves
3M™ Water Filtration Products High Flow Series NEP Valve-In-Head (VH3) with shroud and gauge.
  • ⅜" FNPT, Valve-in-head with Shroud and Gauge
  • FDA CFR-21 compliant materials
  • NSF Certified
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty