Would you like to log in again?
3M™ Zeta Plus™ Single Use Filter Capsule with SP Series Media
Supplier: 3M Healthcare
Zeta Plus™
7100011759 7000051243 7000051626 7010338954 7000051611 7000051238 7000042854 7000051571 7100086226 7000051379 7010382594 7000051256 7000051608 7000051239 7010411141 7010382397 7010303319 7000051605 7000051570 7000125927 7000144869 7000051257 7000051382 7000001846 7010338986 7000051623 7100047617 7010380990 7010415442 7000001845 7100096628 7010338983 7000051258 7100039762 7100009168 7100096294 7010338975 7100116113 7000001839 7010382591 7100029991 7000125922 7010338948 7010380676 7010314168 7010380835 7000051242 7100085896 7000042853 7100069866 7000051586 7000001840 7100031511 7000042856 7100079599 7100029897 7010339897 7100058023 7000051381 7000051254 7100095820 E0170FSA30SP02
76030-836CS 3248.69 USD InStock
76030-836 76033-290 76030-834 76030-832 76030-830 76030-796 76033-258 76030-790 76033-296 76033-254 76251-848 76030-806 76030-804 76030-802 76030-846 76030-800 76030-766 76030-842 76030-764 76030-760 76033-226 76033-260 76031-274 76033-220 76033-266 76033-222 76030-816 76030-814 76030-812 76033-270 76030-854 76030-776 76030-852 76030-850 76253-184 76030-770 76033-238 76242-050 76245-192 76033-272 76033-274 76033-276 76033-232 76033-278 76247-366 76030-828 76030-826 76030-824 76249-010 76030-866 76033-280 76030-788 76030-864 76030-786 76030-862 76030-860 76030-780 76033-248 76033-282 76033-284 76033-242 76033-244
3M™ Zeta Plus™ Single Use Filter Capsule with SP Series Media
Filters Capsule Filters
Zeta Plus™ Encapsulated System Filter Capsule with SP Series Media in the quest for a single-use depth filtration solution that combines a number of ergonomic advantages with the proven performance of our depth filter media. Ergonomic design enables loading and unloading at waist height, resulting in minimal fluid spills when handling spent capsules.
  • FDA Drug Master File (DMF) and USP Class VI Reactivity compliance eases validation and regulatory submission by providing vital documentation and traceability
  • Filter media retains contaminants by mechanical entrapment and electrokinetic adsorption
  • High contaminant holding capacity provides economical filtration and reliable particle reduction
  • Full range of scalable capsule and cartridge filter configurations allow pilot testing and scale-up using the same materials as in full-scale systems
  • Solid core design with fully encapsulated shell eliminates the need for a stainless steel housing and post-filtration cleaning validation
Featuring Zeta Plus™ SP series filter media - Zeta Plus™ Encapsulated System Filter Capsules with SP Media are designed to be used with a Zeta Plus™ Encapsulated System holder and single-use manifolds. Our family of advanced encapsulated depth filters is designed to provide clarification of bioprocess, biological and pharmaceutical fluids. We developed the Zeta Plus™ Encapsulated System Filter Capsule with SP Series filter media to provide optimal clarification of pharmaceutical, biological, bioprocess, and cosmetic fluids.
Our family of Zeta Plus™ SP Series Filter media is composed of advanced cellulose-based depth filtration media, designed to retain contaminants by both mechanical entrapment and electrokinetic adsorption.
Encapsulating high performance - Our encapsulated systems for production scale biomanufacturing (Models# 16EZB and 16EZC) can be pivoted between horizontal and vertical positions. This innovative feature allows for fast and convenient loading and unloading at waist height, minimal footprint during filtration and storage, minimal fluid spills during unloading, and full utilization of the filter media.
In order to facilitate effective monitoring of liquid levels during fill, filtration, and finish, we incorporated a translucent plastic shell on our standard capsules that allows technicians to easily view the liquid level without having to interrupt the process. Another innovative feature is our self-guiding locking mechanism that provides fast and reliable capsule-to-capsule connection.
Applications include Pharmaceuticals, Blood Products, Antibiotics, Cough Syrups, Intravenous Solutions, Growth Media, Bioprocess Fluid and Scalability.
We utilize the effective lenticular filter design and vertical flow path on our Zeta Plus™ Encapsulated System Filter Capsule with SP Media, which has proven effective for depth filtration systems. Combined with our small scale Zeta Plus™ BC filter capsules with SP media, the Zeta Plus™ Encapsulated System Filter Capsules with SP Media offer a complete single-use solution to process from 100 ml to 2500 L of liquid.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty