Zeta Plus™ Encapsulated System Filter Capsule with SP Series Media in the quest for a single-use depth filtration solution that combines a number of ergonomic advantages with the proven performance of our depth filter media. Ergonomic design enables loading and unloading at waist height, resulting in minimal fluid spills when handling spent capsules.
- FDA Drug Master File (DMF) and USP Class VI Reactivity compliance eases validation and regulatory submission by providing vital documentation and traceability
- Filter media retains contaminants by mechanical entrapment and electrokinetic adsorption
- High contaminant holding capacity provides economical filtration and reliable particle reduction
- Full range of scalable capsule and cartridge filter configurations allow pilot testing and scale-up using the same materials as in full-scale systems
- Solid core design with fully encapsulated shell eliminates the need for a stainless steel housing and post-filtration cleaning validation
Featuring Zeta Plus™ SP series filter media - Zeta Plus™ Encapsulated System Filter Capsules with SP Media are designed to be used with a Zeta Plus™ Encapsulated System holder and single-use manifolds. Our family of advanced encapsulated depth filters is designed to provide clarification of bioprocess, biological and pharmaceutical fluids. We developed the Zeta Plus™ Encapsulated System Filter Capsule with SP Series filter media to provide optimal clarification of pharmaceutical, biological, bioprocess, and cosmetic fluids.
Our family of Zeta Plus™ SP Series Filter media is composed of advanced cellulose-based depth filtration media, designed to retain contaminants by both mechanical entrapment and electrokinetic adsorption.
Encapsulating high performance - Our encapsulated systems for production scale biomanufacturing (Models# 16EZB and 16EZC) can be pivoted between horizontal and vertical positions. This innovative feature allows for fast and convenient loading and unloading at waist height, minimal footprint during filtration and storage, minimal fluid spills during unloading, and full utilization of the filter media.
In order to facilitate effective monitoring of liquid levels during fill, filtration, and finish, we incorporated a translucent plastic shell on our standard capsules that allows technicians to easily view the liquid level without having to interrupt the process. Another innovative feature is our self-guiding locking mechanism that provides fast and reliable capsule-to-capsule connection.
Applications include Pharmaceuticals, Blood Products, Antibiotics, Cough Syrups, Intravenous Solutions, Growth Media, Bioprocess Fluid and Scalability.
We utilize the effective lenticular filter design and vertical flow path on our Zeta Plus™ Encapsulated System Filter Capsule with SP Media, which has proven effective for depth filtration systems. Combined with our small scale Zeta Plus™ BC filter capsules with SP media, the Zeta Plus™ Encapsulated System Filter Capsules with SP Media offer a complete single-use solution to process from 100 ml to 2500 L of liquid.