Would you like to log in again?
3M™ Aqua-Pure™ Whole House Scale Inhibition Water Treatment System AP430SS
Supplier: 3M Healthcare
Aqua-Pure™
7100051446 AP43011
76031-259CS 1727.57 USD InStock
76031-259 76031-258
3M™ Aqua-Pure™ Whole House Scale Inhibition Water Treatment System AP430SS
Water Purification Systems Deionizers
We designed the 3M™ Whole House Scale Inhibition Water Treatment System AP430SS to reduce scale build–up on hot water–using appliances such as water heaters (standard and tank–less), clothes washers and dishwashing machines, and on plumbing fixtures like faucets, pipes, and shower heads. The AP430SS provides unrestricted flow of up to 10 gpm with 3/4" NPT inlet/outlet connections.
  • Polyphosphates help to inhibit scale build–up in water heaters (standard and tank–less styles) and in appliances such as dishwashers and clothes washers which can help extend the life of hot water–using appliances
  • Scale inhibition also helps protect piping from corrosion
  • Easy cartridge change–out helps provide easy system maintenance
  • 316 stainless steel head for "sweat–in–place" installation and corrosion resistance
  • FDA CFR–21 compliant materials*
The 3M™ Whole House Scale Inhibition Water Treatment System AP430SS contains polyphosphates to bind calcium and magnesium hardness minerals, preventing them from precipitating out (or becoming a solid) and building up on metallic surfaces such as water heaters and plumbing fixtures like faucets, pipes and shower heads.
Features include: Flow rates up to 10 gpm/37.9 lpm, 25 – 125 psi/172 – 862 kPa pressure range, 40 – 100 °F/37.8 °C temperature range.
The overall design of the scale inhibition system makes the installation and cartridge change–out easy for professionals. The 3M™ Whole House Scale Inhibition Water Treatment System AP430SS is to be installed on the cold–water supply line of the water heater or at point–of–entry of water into the home. The scale inhibition system helps extend the life of downstream water–using appliances such as water heaters (standard and tank–less), clothes washers and dish washing machines, as well as plumbing fixtures like faucets, pipes and shower heads by reducing scale build–up and corrosion. Polyphosphates in minute concentrations of 2–3 ppm, effectively help prevent scale build–up by inhibiting the deposition of hardness minerals onto metallic surfaces. Polyphosphates also helps prevent corrosion by forming a thin protective layer on the metal surface. The 3M™ Scale Inhibition Water Treatment System AP430SS is manufactured using FDA CFR–21 compliant materials* and is suitable for drinking and cooking water, and food applications.
3M branded water filtration products have been addressing water quality issues for over 50 years. 3M products offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Aqua–Pure water quality improvement solutions. 3M, a household brand name, is a known leader in filtration technology recognized for innovation, trust and quality around the world. Having clean and clear drinking water in your home is both a pleasure and a reassurance that you're giving your family the best. With a water filtration system from 3M Purification, you can provide filtered drinking water for every member of your family. From whole home water filtration to drinking water systems, look to 3M Purification to help you find the right solution.
*Surfaces that are in contact with water are FDA CFR–21 compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty