Would you like to log in again?
3M™ Zeta Plus™ BC Series Filter Capsule with ZB Series Media
Supplier: 3M Healthcare
Zeta Plus™
7010301511 7100042717 7100049893 7100032799 7010382845 7100049894 7010296538 7010314487 7010382846 7100049889
76029-561CS 606.39 USD InStock
76029-561 76029-571 76029-563 76029-543 76032-143 76029-533 76029-635 76032-141 76029-645 76029-647
3M™ Zeta Plus™ BC Series Filter Capsule with ZB Series Media
Filters Capsule Filters
Totally disposable capsule depth filters for laboratory, process development, scale-up or small lot production using ZB media.
  • Zeta Plus™ BC units are available in three sizes with industry-leading Zeta Plus™ depth filter media to provide a high degree of application flexibility
  • ZB Grade Cellulose Pre-extracted (acid washed) Diatomaceous Earth (D.E.)30ZB, 60ZB,90ZB
  • Endotoxin reduction in high purity pharmaceutical water systems
The Zeta Plus™ BC Series is a range of completely encapsulated disposable depth filter assemblies, designed for small volume biological, bioprocess and pharmaceutical Filtration.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty