Would you like to log in again?
3M™ NanoSHIELD™ MDC Series Hollow Fiber Filter Capsule
Supplier: 3M Healthcare
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NanoSHIELD™
7010301485 7010382306 7010303343 7010339218 7010314519 7010410036 7010381195 7010413620 7000125745 7010338886 7010340147 7010315730 7010382994 7010339260 7010383132 7010339971
76032-624CS 1455.16 USD InStock
76032-624 76032-558 76032-622 76032-544 76032-606 76032-604 76032-548 76032-582 76033-738 76032-570 76032-580 76032-620 76032-554 76032-542 76032-618 76033-740
3M™ NanoSHIELD™ MDC Series Hollow Fiber Filter Capsule
Filters Capsule Filters
NanoSHIELD hollow fiber series family of products is offered in several configurations to allow the end user to match the volume of fluid with the appropriate filter in an effort to reduce chemical waste.
  • Small, compact design make MDC capsules
  • Ideal for applications requiring low hold-up volume
  • High particle rentention from 100 nm down to 5 nm
MDC Capsules for advanced photoresists and chemicals applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty