Would you like to log in again?
3M™ LifeASSURE™ PDA Series Filter Capsule
Supplier: 3M Healthcare
LifeASSURE™
7000051537 7100017245 7000051529 7000051535 7000051532 7000051533 7000051536 7000051530 7100087472 7000051531 7100011132 7000051534 7100005500 7000051527
76034-002CS 844.07 USD InStock
76034-002 76032-744 76034-000 76032-742 76028-498 76032-746 76034-018 76034-006 76034-016 76032-740 76034-004 76033-994 76033-996 76033-998
3M™ LifeASSURE™ PDA Series Filter Capsule
Filters Capsule Filters
This capsule made of two asymmetric polyethersulfone (PES) membrane layers with our Advanced Pleat Technology in manufacturing LifeASSURE™ PDA Series Filter Capsule. This creates a filter with high throughput and fast flow. These filters have applications in pre-filtration and sterilizing filtration in the pharmaceutical, biological, and bioprocess industries where low protein binding is critical.
  • Validated 0.2 and 0.1 µm rated membrane provides reliable sterilizing filtration
  • Excellent contaminant capacity, flow rates, and life resulting from robust double-asymmetric membrane layer filter construction
  • Hydrophilic asymmetric polyethersulfone membrane enables wet out in aqueous solutions for integrity testing
  • Advanced Pleat Technology construction increases the useful surface area by increasing the flow paths between media pleats
  • 100% forward flow Integrity tested prior to release
  • Both pre-sterilized and sterilizable LifeASSURE™ PDA capsules are available
Engineered for Quality and Reliability.
The 3M Purification Inc. Quality Management System is approved by an accredited registering body to the ISO 9001:2008 and ISO 13485:2003 Quality Management Systems, and the cartridges are 100% forward flow integrity tested prior to shipment. We ship all LifeASSURE™ PDA cartridge and capsule filters with a Certificate of Quality affirming compliance with manufacturing quality specifications. Additionally, supporting Drug Master File (DMF) documentation is on file with the United States Food and Drug Administration (FDA) and a complete LifeASSURE PDA Validation Guide is available upon request.
Meeting Your Scale-Up Needs.
We are pleased to offer a full range of LifeASSURE™ PDA sterilizing-grade filters to meet your scale-up process needs. This includes pre-sterilized or sterilizable disposable capsules in various sizes as well as full-size cartridges. We also offer filter discs and 50 mm capsules when even smaller membrane surface areas are required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty