Would you like to log in again?
3M™ AL Series Filter Housing
Supplier: 3M Healthcare
3M™
7010338619 7100054435 7000125596 7000125561
76034-550CS 5929.71 USD InStock
76034-550 76031-810 76034-548 76031-808
3M™ AL Series Filter Housing
Filters Filter Cartridges
3M™ AL Series Filter Housing is constructed of durable, cost–effective cast iron and steel to create a quality–built vessel on the outside, designed to satisfy a wide variety of general–purpose industrial and commercial filtration needs. On the inside, long–lasting stainless steel internal components resist corrosion and help make cleaning and maintenance quick and easy.
  • Durable cast iron and steel with stainless steel internals
  • Housings will accept modular 9³/₄" and 10" length cartridges
  • Smart, 3–piece construction disassembles for quick and easy cleaning
  • Mounting pads securely hold housing in place
  • Convenient drains and petcock vent
It is designed to accommodate liquids at a maximum of 150 psi at 250 °F in a wide range of sizes, from a 3 cartridge model to an 18 cartridge model.
Features a three piece design with cartridge centerpost construction. As process requirements evolve and grow and as floor plans change, you will appreciate the sensibility and convenience of our three–piece design when you need to assemble, disassemble, clean and perform periodic maintenance. Our cartridge centerpost construction holds the filters in place and helps make cartridge change–outs quick and easy. The cartridge posts are removable to help make cleaning and maintenance a time and cost–saving endeavor.
Applicable filter cartridges that can be used in the 3M™ AL Housing Series include the following: Betapure AU™ Series Filters, Betapure™ NT–T Series Filters, Micro–Klean™ RB Series Filters, Micro–Klean™ CS Series Filters and Micro–Klean™ RT Series Filters.
The AL housing can be used in systems with flow rates up to 105 gpm (for water).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty