Would you like to log in again?
Cell-Mate3D™ Cell Retrieval Kit, BRTI Life Sciences
Supplier: BRTI Life Sciences
RET-1001
76036-026PK 74.42 USD InStock
76036-026
Cell-Mate3D™ Cell Retrieval Kit, BRTI Life Sciences
Assays Cellular Assays
Isolate cells from Cell-Mate3D matrix simple 15 minutes process cancer, stem cells, normal cells.
  • Up to 85% recovery of viable cells
  • Simple and easy 15 minutes matrix digestion
  • Use for flow cytrometry
  • Sort specific cell populations
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty