Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Tonbo Biosciences
35-8065-M002
76040-194EA 273 USD InStock
76040-194
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat anti-Mouse IgM (mu chain) - Affinity Pure, FITC Conjugate

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty