Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgM µRabbit Polyclonal Antibody (Biotin)

Supplier: Tonbo Biosciences
30-8075-M001
76040-212EA 429 USD InStock
76040-212
Anti-IgM µRabbit Polyclonal Antibody (Biotin)
Antibodies
Rabbit anti-Human IgM (mu chain) - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: Human IgM (µ chain)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ chain
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty