Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG2b Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: Tonbo Biosciences
50-8035-U100
76040-236EA 313 USD InStock
76040-236
Anti-IgG2b Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Goat anti-Mouse IgG2b, secondary antibody, conjugated to SureLight® R-Phycoerythrin

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty