Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD45 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cyanine5) [clone: 2D1]

Supplier: Tonbo Biosciences
55-9459-T025 55-9459-T100
76040-294EA 276 USD InStock
76040-294 76040-292
Anti-CD45 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cyanine5) [clone: 2D1]
Antibodies
The 2D1 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, and CD45RO. The 2D1 antibody is widely used as a marker for human CD45 expression on peripheral blood T cells, B cells, monocytes, macrophages, and NK cells.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 2D1
Conjugation: PE (Phycoerythrin)-Cyanine5
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty