Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD16 / CD32 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cyanine7) [clone: 2.4G2]

Supplier: Tonbo Biosciences
60-0161-U025 60-0161-U100
76040-306EA 66.9 USD InStock
76040-306 76040-308
Anti-CD16 / CD32 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cyanine7) [clone: 2.4G2]
Antibodies
The 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). These receptors are expressed by B cells, monocytes, macrophages, NK cells, dendritic cells, and neutrophils.

Type: Primary
Antigen: CD16 / CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: PE (Phycoerythrin)-Cyanine7
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty