Would you like to log in again?
PLATE SYSTEM RACK U2D CHEM RESISTANT MAT
Supplier: MicroSolv Technology Corp
Easy Purchase Pack U-2D* Plate System Rack with 96 1ML insert^ Easy Purchase Pack, U-2D* Plate System Rack with 96 1ML inserts made with RSA-Pro glass preloaded in rack. Includes base & Chemical resistant sealing mat, Non-slit
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE SYSTEM RACK U2D CHEM RESISTANT MAT 76040-522 95296-1ML-1RMR Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty