Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Non Sterile Carbon Steel Surgical Blades, Propper Manufacturing
Supplier: Propper
12102200 12102000 12101000 12101500 12101100
76044-624BX 76.15 USD InStock
76044-624 76044-626 76044-638 76044-640 76044-628
Non Sterile Carbon Steel Surgical Blades, Propper Manufacturing
Scalpels and Scalpel Assemblies Scalpel Blades
Propper non-sterile carbon steel surgical blades are manufactured with razor-sharp cutting edges allowing for precision and clean incisions.
  • Superior cutting ability of carbon steel
  • Non-sterile blades
  • Enclosed in heat-sealed peel-pack
Non-sterile carbon steel blades offer the superior cutting advantages of carbon steel technology. Available in all necessary sizes. Blades feature consistent sharpness and strength required for the most demanding procedures.
Description of use of each size
#10: Curved cutting edge is one of the more traditional blade shapes, making small incisions to the skin and muscle clean and precise.
#11: Elongated triangular blade with a strong tip, ideal for stab incisions. Most commonly used for the incisions of chest drains, and opening various coronary arteries.
#15: Small curved cutting edge is the most popular blade for short and precise incisions.
#20: A larger version of the No. 10 blade equipped with a curved cutting edge, used mostly in orthopedic and general surgical procedures.
#22: A tad bigger than the number #21 blade, used for skin incisions in both cardiac and thoracic surgery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty