Would you like to log in again?
Fine Surgical Blades, Propper Manufacturing
Supplier: Propper
12706100 12706700 12706400 12706200
76044-664BX 124.97 USD InStock
76044-664 76044-666 76044-660 76044-662
Fine Surgical Blades, Propper Manufacturing
Scalpels and Scalpel Assemblies Scalpel Blades
Propper fine surgical blades are small, slim precise blades for your most delicate procedures.
  • Sterile fine surgical blades
  • For use with any chuck handle
Propper Fine surgical blades are designed for microsurgical procedures including reconstruction, ophthalmic and cardiology. Blades wrapped individually in peel-pack sterile foil pouches. Propper Fine Surgical Blades are not made with rubber latex.
Size Details
#61: A chisel shaped blade used for nail treatments in podiatry and chiropody.
#62: A slightly wider chisel shape used for nail treatments in podiatry and chiropody.
#64: Can be used in a number of specialist disciplines, ideal for plastic and reconstructive surgery, hip and knee arthroscopy and tenotomy.
#67: Can be used in a variety of specialties including plastic and reconstructive surgery, ophthalmology cardiovascular surgery, and dental periodontics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty