Would you like to log in again?
DATA LOGER LIBERO WITH WSI SENSOR ONLY
Supplier: ELPRO SERVICES, INC.
Data Loger LIBERO^ Data Loger LIBERO, with WSI sensor only, for Refrigerator Monitoring Exchangeable and calibrated puck sensor only (-10C. +50C)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DATA LOGER LIBERO WITH WSI SENSOR ONLY 76045-178 801416 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty