Would you like to log in again?
WIPER SEALED EDGE CLEANROOM PES 12X12IN
Supplier: Texwipe
Sealed Edge Polyester Cleanroom Wiper 12in X 12in^ Sealed Edge Polyester Cleanroom Wiper, Dimensions: 12in X 12, Good For Spill Control And General Cleaning Applications, Safe For Use With Many Solvents Including Ipa, Used In Iso Class 4 And Higher Enviro
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WIPER SEALED EDGE CLEANROOM PES 12X12IN 76045-364 TX8942 Bag of 100 Retrieving
Case of 1000 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty