Would you like to log in again?
WIPER LAUNDERED MICROFIBER SEALED 9X9IN
Supplier: Texwipe
Laundered Microfiber Wiper 9X9in^ Laundered Microfiber Wiper, Sealed Edge, Dimensions: 9X9in, 80% Pet / 20% Nylon Wedge Fiber Wipes Surfaces Dry No Streaking Iso 3 Environment, Ideal For Cleaning Delicate Sensitive Surfaces Such As Optics Removes Finger P
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WIPER LAUNDERED MICROFIBER SEALED 9X9IN 76045-376 TX1209 Bag of 100 (1/Itm/Bx) Retrieving
Case of 1000 (1/ Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty