Would you like to log in again?
Low Temperature PCR Racks
Supplier: Heathrow Scientific
HS120540 120539 HS120541 HS120538
76045-412CS 421.96 USD InStock
76045-412 76045-416 76045-414 76045-408
Low Temperature PCR Racks
Racks Tube Racks
These Low Temperature PCR Racks feature a sliding lid that roates 180° to become a convenient stand that the rack sits on. These PCR racks also feature ergonomic textured finger grips that provide a secure hold, reducing the chance of dropping the rack, and makes handling and transportation easy.
  • Polycarbonate lid serves as cover and elevated rack base
  • Lid can be used in right or left-handed orientation and is detachable
  • Sliding lid rotates 180° to become a convenient stand that the rack sits on
  • Side opening enables faster cooling of samples to ensure integrity
  • Stackable design saves valuable bench-top and freezer space
  • Features alphanumeric identification for quick location and retrieval of tubes
  • Can be used for freezer storage down to −80 °C
The lids have a side opening that enables faster cooling of samples to ensure integrity and are made from translucent polycarbonate for easy sample and well identification. The base of the rack is made from robust ABS construction that helps keep samples safe, so you don't have to spend extra time and money on expensive retesting. This rack comes in multiple colors for coding options in the lab.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty