Would you like to log in again?
LaboPlast® Bürkle Thread Adapter, 3/4" Inflowing, Coarse Thread , Tri-Sure 2" Outflowing, Bürkle
Supplier: Bürkle
0600-0094
76045-418EA 31.66 USD InStock
76045-418
LaboPlast® Bürkle Thread Adapter, 3/4" Inflowing, Coarse Thread , Tri-Sure 2" Outflowing, Bürkle
Fittings and Connectors
Designed for use with pumps, these adapters feature a coarse, Tri-Sure 2" thread on the outflowing side, and a ³/₄" thread on the inflowing side.
Stopcock spigots, ball valves, and other accessories can be connected to most commercially available containers with LaboPlast® thread adapters.
Packaging: One adapter per package.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty