Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brilliant green solution 1% (w/v) Counterstain

Supplier: HARDY DIAGNOSTICS
Synonyms: CI 42040Basic Green 1N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulfateBrilliant Green hydrogen sulfateBrilliant Green hydrogen sulphateDiamond GreenAstradiamant green GXN-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphateSolid Green JOEthyl GreenMalachite Green GEmerald Green

BG008
76045-452EA 18.41 USD InStock
76045-452
Brilliant green solution 1% (w/v) Counterstain
Brilliant Green
For acid-fast stain procedures and other uses

1% weight/volume. A saline solution of brilliant green.


Formula: C₂₇H₃₄N₂O₄S
MW: 482.65 g/mol
MDL Number: MFCD00011880
CAS Number: 633-03-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty