Would you like to log in again?
Tryptic Soy Broth (TSB) with Lecithin and Tween®
Supplier: Hardy Diagnostics
U134G U306 U292 U134G U229 U425 U261 U241 U423 U424 K134 K134
89408-126PK 399.06 USD InStock
89408-126 10135-882 10124-508 10124-506 10053-676 10135-886 10053-678 10127-494 76045-460 CA10127-494 10135-878 CA76047-198
Tryptic Soy Broth (TSB) with Lecithin and Tween®
Media for Microbiology Dehydrated Media for Microbiology
Recommended for use in the cultivation of a wide variety of fastidious and nonfastidious microorganisms.
  • For fastidious and nonfastidious microorganisms
  • With neutralizing additives
Lecithin and Tween® 80 help in neutralizing residual sanitizing chemicals and help in emulsifying samples. This product is not intended for use in diagnosing human disease.
Ordering information: This product is not intended to be used for the diagnosis of human disease.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty