Would you like to log in again?
Purple Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Y115 Y115 Y114 Y108
76045-468PK 874.92 USD InStock
76045-468 CA76047-208 76045-470 76045-442
Purple Broth, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the determination of carbohydrate fermentation by bacteria, especially enteric bacilli and Enterococcus spp
Contains a durham tube for gas detection.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty