Would you like to log in again?
Human Recombinant Growth and differentiation factor-5
Supplier: Enzo Life Sciences
201-802-0010 201-802-0100 201-802-0050
76045-522EA 332.69 USD InStock
76045-522 76045-520 76288-318
Human Recombinant Growth and differentiation factor-5
Proteins and Peptides
  • High purity
  • Carrier-free
Produced in E. coli. Non-glycosylated homodimer, containing two 117 amino acids chains. To enable bacterial expression the N-terminal sequence of Ala-Pro-Leu-Thr was replaced with a Lys.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty