Would you like to log in again?
ADAPTER SET BLOOD BAG 3600 RPM SMALL
Supplier: THERMO EC
Adapter for Small Blood Bag and Cell Culture Bag^ Adapter for Small Blood Bag and Cell Culture Bag, Maximum Speed: 3,600 RPM, Maximum RCF: 3,028 x g, 2 places/adapter
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER SET BLOOD BAG 3600 RPM SMALL 76045-586 75003829 Pack of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty