Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cole-Parmer® SHM-250 Series Digi-Mantle Heating Kits, Antylia Scientific

Supplier: Antylia Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Cole-Parmer®
36204-21 36204-25 36204-27 36204-11 36204-17
76045-874EA 5073.62 USD InStock
76045-874 76045-878 47742-566 76045-876 76045-880
Cole-Parmer® SHM-250 Series Digi-Mantle Heating Kits, Antylia Scientific
Heaters Heating Mantles
Heating kit includes a heating mantle controller and interchangeable heating cartridge.

  • Achieve uniform temperature up to 450 °C across round bottom flask
  • Stirring speeds from 100 to 2000 rpm with excellent stability
  • Vented casing keeps heating cartridge cool to the touch
  • Modular design allows different size heating cartridges to be used on the same Digi-Mantle controller

Control temperature and stirring speed independently on the Digi-Mantle digital controller. The control panel features indicator lights for power, heater, and stirrer. Microprocessor-controlled heater displays the heat setting as a percentage of the total power. View the actual temperature (up to 220 °C) of the flask content on a three digit LCD display by using the included temperature probe.

Each digi-mantle heating kit includes a controller and a heating cartridge that accommodates 100 to 250 ml, 500 or 1000 ml round bottom flasks. Due to its modular design a different size heating cartridge can be used with the same controller, saving money.

Ordering information: Supplied with a support rod. Clamps are not supplied.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty