Would you like to log in again?
Microbalance, WXTS University Edition, METTLER TOLEDO®
Supplier: Mettler-Toledo
30376748
76046-386EA 24350 USD InStock
76046-386
Microbalance, WXTS University Edition, METTLER TOLEDO®
Balances and Scales
The WXTS3DU microbalance is a great choice for academic research and testing. The balance offers a rugged and durable design with simple operation. With a capacity up to 1.2 / 3.2 g (fine / full range) and readability from 0.0001 / 0.01 mg this balance is equipped to deliver consistent and fast results every time.
  • Intuitive touchscreen operation
  • Robust aluminum housing
  • Easy and efficient cleaning
  • Durable magnetic force restoration weighing sensor
  • FACT - Fully Automatic Calibration Technology
  • Internal adjustment
  • Two AUX Ports for external switches
  • Integrated RS232 interface
  • Slot for optional RS, Ethernet or Bluetooth
  • Centrally placed level bubble
  • Data transfer key to PC host or system
The WXTS3DU is a great solution for academic research and testing. The successor to the XS3DU Microbalance is a durable solution that will offer reliable results for many years to come.
The balance has minimum sample weight (load = 200 mg, k=2, U=1%) of 0.14 mg and the USP minimum sample weight (load = 200 mg, k=2, U=0.10%) is 1.4 mg.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty