Would you like to log in again?
BRIGHTSORB™ Heavy Weight Chemical Absorbent Roll, Brady Worldwide
Supplier: Brady
107691
76046-568RL 432.12 USD InStock
76046-568
BRIGHTSORB™ Heavy Weight Chemical Absorbent Roll, Brady Worldwide
Spill Control Materials
BRIGHTSORB™ High Visibility absorbents feature bright safety yellow coloring for easy separation of hazardous waste
  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Heavy weight design specifically intended to use less material for continuous leaks and larger spills
  • BRIGHTSORB™ High Visibility series combine safety signage and absorbency in one product
  • Perforations provide cost control and reduce waste by allowing use of only the amount needed
Greater performance without the extra weight. Specially developed heavy weight pads feature a chemical sorbent and are yellow color-coded for safety and easy separation of hazardous waste. Material does not react with aggressive fluids. Perforated every 15", and up the center.
Ideal in areas with a high risk of slips, trips, and falls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty