Would you like to log in again?
LENS ANTI-FOG SCOTCHGARD PLYCARBO AMBER
Supplier: 3M OCCUPATIONAL HEALTH
SecureFit* Scotchgard* Anti-fog lens Amber^ SecureFit* Scotchgard* Anti-fog lens, Eyewear Range Premium, Frame Color: Gray, Frame Material: Plastic, Frame Style/Type: Half-frame, Lens Color: Amber, Lens Marking: Z87.1+, Lens Material: Polycarbonate, ANSI Z87.1-2003, UV Protection 99.9%
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LENS ANTI-FOG SCOTCHGARD PLYCARBO AMBER 76046-608 7100141599 Case of 20 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty