Would you like to log in again?
Human Recombinant GH (from E. coli)
Supplier: Shenandoah Biotechnology
100-40AF-1MG 100-40AF-5MGS 100-40AF-10MGS 100-40AF-100UG 100-40AF-500UG
76046-762EA 394.56 USD InStock
76046-762 76046-764 76046-766 76046-758 76046-760
Human Recombinant GH (from E. coli)
Proteins and Peptides
Growth hormone (GH) is an important mitogenic growth factor that is synthesized, stored, and secreted by somatotropic cells of the anterior pituitary gland. GH stimulates growth, cell reproduction, and cell regeneration. In children, GH deficiencies can cause short stature, growth failure, and delayed sexual maturity. Adult GH deficiency presents with reduced lean body mass, increased adiposity, reduced muscle strength, and ultimately premature mortality. GH replacement therapy is used to treat many growth disorders, including Turner syndrome, chronic renal failure, and Prader–Willi syndrome.
  • US-made recombinant proteins
  • Strict quality standards
  • Low manufacturer pricing
Growth hormone (GH) is an important mitogenic growth factor that is synthesized, stored, and secreted by somatotropic cells of the anterior pituitary gland. GH stimulates growth, cell reproduction, and cell regeneration. In children, GH deficiencies can cause short stature, growth failure, and delayed sexual maturity. Adult GH deficiency presents with reduced lean body mass, increased adiposity, reduced muscle strength, and ultimately premature mortality. GH replacement therapy is used to treat many growth disorders, including Turner syndrome, chronic renal failure, and Prader–Willi syndrome. 
Certifications: Animal-Free (AF) proteins are identical to regular proteins and offer additional documentation showing that they were produced with no materials of animal or human origin using ANIMAL-FREE purification processes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty