Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lyovapor™ L-200 Pro Control Freeze Dryers, Büchi
Supplier: Büchi
11L200PH2422111002 200PH242111002 11L200PH2432111002 11L200PH2432111002 11L200PH2422111002 200PH243111002 200PH242111002 200PH243111002
76046-840EA 45708.16 USD InStock
76046-840 CA76046-840 76046-834 CA76046-834 76046-836 CA76046-836 76046-838 CA76046-838
Lyovapor™ L-200 Pro Control Freeze Dryers, Büchi
Freeze Dryers
The compact Lyovapor™ L-200 offers high-quality Freeze Drying with complete automation.
  • Fully automated
  • Method creation and reporting
  • Real-time pressure and temperature recording
  • Stoppering available under inert atmospheric conditions
  • Small footprint
  • Touch-screen control panel
The Pro Control L-200 Lyovapor™ offers the highest quality freeze drying with complete automation. A 6L condenser capacity allows for maximum sample throughput. Integrating Infinite-Control™ with your freeze dryer gives you access to all process parameters and keeps you informed by push notifications anywhere, anytime.
Certifications: CE/CSA certified.
Ordering information: The Pfieffer Duo 6 vacuum pump is required for automation. Additional sampling platforms are available upon inquiry. Additional configurations are available. Contact customer service for further details.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty