Would you like to log in again?
Extracta™ DNA Prep for PCR, Quantabio
Supplier: Quantabio
Extracta™
95091-250 95091-025 95091-002 95091-025 95091-250 95091-002
76047-142EA 23.4 USD InStock
76047-142 CA97065-352 CA97065-350 97065-352 97065-350 CA76047-142
Extracta™ DNA Prep for PCR, Quantabio
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Extracta DNA Prep for PCR is a two-component reagent kit for rapid extraction of PCR-ready genomic DNA from a variety of tissues. Samples are processed in less than 30 minutes with minimal hands-on time and technical skill.
  • Simple, fast protocol
  • Compatible with laboratory automation
  • DNA is suitable for qPCR, High Resolution Melt Analysis, endpoint PCR and other applications
  • Works with wide range of tissue including hair, buccal swabs and mouse tails
  • Optional 10 minute protocol for high-throughput applications
Quanta's Extracta sample preparation system is a system that can be scaled to process various amounts of solid tissue in a simple and fast single-tube procedure. DNA may be purified from many tissue types and is suitable for use in sensitive downstream applications such as PCR, qPCR and High Resolution Melt Analysis.
The Extracta™ DNA prep for PCR consists of a two component reagent kit for rapid and efficient extraction of PCR-ready genomic DNA from mammalian tissues. The kit has been demonstrated to work with mouse tail biopsies, ear punches and a variety of human tissue samples including hair, buccal cells and saliva. The genomic DNA is suitable for sensitive downstream PCR applications including end-point PCR, High Resolution Melt Analysis (HRM) and quantitative real time PCR (qPCR) without requiring any additional clean-up. In addition, the extracted DNA may be used in multiplexed PCR applications such as transgene or knock-out analyses. Tissue extractions can be performed in tubes, plates or deep well blocks to allow for adaptation to workflow and automation on liquid handling workstations.
Ordering information: DNA Prep kit includes extraction reagent and stabilization buffer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty